Anti-TMEM115 Mouse Monoclonal Antibody [clone: 3D4]

Supplier: Abnova
H00011070-M01
10597-176EA 335 USD
10597-176
Anti-TMEM115 Mouse Monoclonal Antibody [clone: 3D4]
Antibodies
Type: Primary
Antigen: TMEM115
Clonality: Monoclonal
Clone: 3D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR