Anti-UBE2C Mouse Monoclonal Antibody [clone: 9D3]

Supplier: Abnova
H00011065-M01
10605-216EA 335 USD
10605-216
Anti-UBE2C Mouse Monoclonal Antibody [clone: 9D3]
Antibodies
Type: Primary
Antigen: UBE2C
Clonality: Monoclonal
Clone: 9D3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR