Anti-SOX30 Mouse Monoclonal Antibody [clone: 6H1]

Supplier: Abnova
H00011063-M02
10601-784EA 335 USD
10601-784
Anti-SOX30 Mouse Monoclonal Antibody [clone: 6H1]
Antibodies
Type: Primary
Antigen: SOX30
Clonality: Monoclonal
Clone: 6H1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR