Anti-NUDT21 Mouse Monoclonal Antibody [clone: 3F8]

Supplier: Abnova
H00011051-M12
10582-822EA 335 USD
10582-822
Anti-NUDT21 Mouse Monoclonal Antibody [clone: 3F8]
Antibodies
Type: Primary
Antigen: NUDT21
Clonality: Monoclonal
Clone: 3F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR