Anti-NUDT21 Mouse Monoclonal Antibody [clone: 2G4-6F11]

Supplier: Abnova
H00011051-M01A
10624-110EA 335 USD
10624-110
Anti-NUDT21 Mouse Monoclonal Antibody [clone: 2G4-6F11]
Antibodies
Type: Primary
Antigen: NUDT21
Clonality: Monoclonal
Clone: 2G4-6F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR