Anti-SLC35D2 Mouse Monoclonal Antibody [clone: 1H5]

Supplier: Abnova
H00011046-M03
10601-636EA 335 USD
10601-636
Anti-SLC35D2 Mouse Monoclonal Antibody [clone: 1H5]
Antibodies
Type: Primary
Antigen: SLC35D2
Clonality: Monoclonal
Clone: 1H5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR