Anti-SBLF Mouse Monoclonal Antibody [clone: 1F3]

Supplier: Abnova
H00011037-M01
10600-676EA 335 USD
10600-676
Anti-SBLF Mouse Monoclonal Antibody [clone: 1F3]
Antibodies
Type: Primary
Antigen: SBLF
Clonality: Monoclonal
Clone: 1F3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR