Anti-GTF2A1L Mouse Polyclonal Antibody

Supplier: Abnova
H00011036-B01
10510-728EA 325 USD
10510-728
Anti-GTF2A1L Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GTF2A1L (general transcription factor IIA, 1-like)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR