Anti-RAB31 Mouse Monoclonal Antibody [clone: 4D12]

Supplier: Abnova
H00011031-M03
10598-868EA 335 USD
10598-868
Anti-RAB31 Mouse Monoclonal Antibody [clone: 4D12]
Antibodies
Type: Primary
Antigen: RAB31
Clonality: Monoclonal
Clone: 4D12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR