Anti-IL24 Rabbit Polyclonal Antibody

Supplier: Abnova
H00011009-D01P
10590-290EA 376 USD
10590-290
Anti-IL24 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL24
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR