Anti-RAB32 Mouse Polyclonal Antibody

Supplier: Abnova
H00010981-B01P
10509-796EA 335 USD
10509-796
Anti-RAB32 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RAB32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR