Anti-SF3A3 Mouse Monoclonal Antibody [clone: 2E9-2B7]

Supplier: Abnova
H00010946-M01A
10625-088EA 335 USD
10625-088
Anti-SF3A3 Mouse Monoclonal Antibody [clone: 2E9-2B7]
Antibodies
Type: Primary
Antigen: SF3A3
Clonality: Monoclonal
Clone: 2E9-2B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR