Anti-MSL3 Mouse Polyclonal Antibody

Supplier: Abnova
H00010943-B01P
10564-210EA 335 USD
10564-210
Anti-MSL3 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MSL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR