Anti-RALBP1 Mouse Monoclonal Antibody [clone: 1C2-A10]

Supplier: Abnova
H00010928-M01
10599-108EA 335 USD
10599-108
Anti-RALBP1 Mouse Monoclonal Antibody [clone: 1C2-A10]
Antibodies
Type: Primary
Antigen: RALBP1
Clonality: Monoclonal
Clone: 1C2-A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR