Anti-SUB1 Mouse Monoclonal Antibody [clone: 2D6]

Supplier: Abnova
H00010923-M13
10596-152EA 335 USD
10596-152
Anti-SUB1 Mouse Monoclonal Antibody [clone: 2D6]
Antibodies
Type: Primary
Antigen: SUB1
Clonality: Monoclonal
Clone: 2D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR