Anti-MAGED2 Mouse Monoclonal Antibody [clone: 6G10]

Supplier: Abnova
H00010916-M04
10592-482EA 335 USD
10592-482
Anti-MAGED2 Mouse Monoclonal Antibody [clone: 6G10]
Antibodies
Type: Primary
Antigen: MAGED2
Clonality: Monoclonal
Clone: 6G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR