Anti-EDAR Mouse Monoclonal Antibody [clone: 6C12]

Supplier: Abnova
H00010913-M01
10584-568EA 335 USD
10584-568
Anti-EDAR Mouse Monoclonal Antibody [clone: 6C12]
Antibodies
Type: Primary
Antigen: EDAR
Clonality: Monoclonal
Clone: 6C12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR