Anti-SUGT1 Mouse Monoclonal Antibody [clone: 1A10]

Supplier: Abnova
H00010910-M04
10602-926EA 335 USD
10602-926
Anti-SUGT1 Mouse Monoclonal Antibody [clone: 1A10]
Antibodies
Type: Primary
Antigen: SUGT1
Clonality: Monoclonal
Clone: 1A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR