Anti-SUGT1 Mouse Monoclonal Antibody [clone: 6G5]

Supplier: Abnova
H00010910-M03
10602-924EA 335 USD
10602-924
Anti-SUGT1 Mouse Monoclonal Antibody [clone: 6G5]
Antibodies
Type: Primary
Antigen: SUGT1
Clonality: Monoclonal
Clone: 6G5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR