Anti-CPSF4 Mouse Polyclonal Antibody

Supplier: Abnova
H00010898-B01
10553-750EA 335 USD
10553-750
Anti-CPSF4 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CPSF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR