Anti-PPARGC1A Mouse Monoclonal Antibody [clone: 2F9]

Supplier: Abnova
H00010891-M04
10597-724EA 335 USD
10597-724
Anti-PPARGC1A Mouse Monoclonal Antibody [clone: 2F9]
Antibodies
Type: Primary
Antigen: PPARGC1A
Clonality: Monoclonal
Clone: 2F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR