Anti-NMU Rabbit Polyclonal Antibody

Supplier: Abnova
H00010874-D01P
10595-058EA 376 USD
10595-058
Anti-NMU Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: NMU
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR