Anti-HCP5 Rabbit Polyclonal Antibody

Supplier: Abnova
H00010866-D01P
10587-840EA 376 USD
10587-840
Anti-HCP5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HCP5 (HLA complex P5 (non-protein coding))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR