Anti-HCP5 Mouse Polyclonal Antibody

Supplier: Abnova
H00010866-B01P
10558-162EA 335 USD
10558-162
Anti-HCP5 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HCP5 (HLA complex P5 (non-protein coding))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR