Anti-CYP46A1 Mouse Monoclonal Antibody [clone: 2B5]

Supplier: Abnova
H00010858-M02
10582-540EA 335 USD
10582-540
Anti-CYP46A1 Mouse Monoclonal Antibody [clone: 2B5]
Antibodies
Type: Primary
Antigen: CYP46A1
Clonality: Monoclonal
Clone: 2B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR