Anti-FTCD Mouse Monoclonal Antibody [clone: 5F6]

Supplier: Abnova
H00010841-M02
10586-380EA 335 USD
10586-380
Anti-FTCD Mouse Monoclonal Antibody [clone: 5F6]
Antibodies
Type: Primary
Antigen: FTCD (formimidoyltransferase cyclodeaminase)
Clonality: Monoclonal
Clone: 5F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR