Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: 3E9]

Supplier: Abnova
H00010840-M01
10510-678EA 335 USD
10510-678
Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: 3E9]
Antibodies
Type: Primary
Antigen: ALDH1L1
Clonality: Monoclonal
Clone: 3E9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR