Anti-FRS2 Mouse Monoclonal Antibody [clone: 1F7-1D6]

Supplier: Abnova
H00010818-M02
10586-710EA 335 USD
10586-710
Anti-FRS2 Mouse Monoclonal Antibody [clone: 1F7-1D6]
Antibodies
Type: Primary
Antigen: FRS2 (fibroblast growth factor receptor substrate 2)
Clonality: Monoclonal
Clone: 1F7-1D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR