Anti-FRS2 Mouse Polyclonal Antibody

Supplier: Abnova
H00010818-A02
10557-070EA 243 USD
10557-070
Anti-FRS2 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FRS2 (fibroblast growth factor receptor substrate 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR