Anti-NEK6 Mouse Monoclonal Antibody [clone: 3B5]

Supplier: Abnova
H00010783-M01
10594-676EA 335 USD
10594-676
Anti-NEK6 Mouse Monoclonal Antibody [clone: 3B5]
Antibodies
Type: Primary
Antigen: NEK6 (NIMA-related kinase 6)
Clonality: Monoclonal
Clone: 3B5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR