Anti-ZNF274 Mouse Monoclonal Antibody [clone: 2F5]

Supplier: Abnova
H00010782-M02
10606-428EA 335 USD
10606-428
Anti-ZNF274 Mouse Monoclonal Antibody [clone: 2F5]
Antibodies
Type: Primary
Antigen: ZNF274
Clonality: Monoclonal
Clone: 2F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR