Anti-HBS1L Rabbit Polyclonal Antibody

Supplier: Abnova
H00010767-D01
10588-360EA 376 USD
10588-360
Anti-HBS1L Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HBS1L (HBS1-like (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR