Anti-KDM5B Mouse Monoclonal Antibody [clone: 1G10]

Supplier: Abnova
H00010765-M02
10590-974EA 335 USD
10590-974
Anti-KDM5B Mouse Monoclonal Antibody [clone: 1G10]
Antibodies
Type: Primary
Antigen: KDM5B
Clonality: Monoclonal
Clone: 1G10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR