Anti-TRAF3IP2 Mouse Monoclonal Antibody [clone: 4A3]

Supplier: Abnova
H00010758-M01
10581-268EA 335 USD
10581-268
Anti-TRAF3IP2 Mouse Monoclonal Antibody [clone: 4A3]
Antibodies
Type: Primary
Antigen: TRAF3IP2
Clonality: Monoclonal
Clone: 4A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR