Anti-CERS1 Mouse Polyclonal Antibody

Supplier: Abnova
H00010715-A01
10562-852EA 243 USD
10562-852
Anti-CERS1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CERS1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR