Anti-CCT6B Mouse Monoclonal Antibody [clone: 1A4]

Supplier: Abnova
H00010693-M01
10578-996EA 335 USD
10578-996
Anti-CCT6B Mouse Monoclonal Antibody [clone: 1A4]
Antibodies
Type: Primary
Antigen: CCT6B
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR