Anti-CCT6B Mouse Polyclonal Antibody

Supplier: Abnova
H00010693-B01P
10560-168EA 335 USD
10560-168
Anti-CCT6B Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCT6B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR