Anti-GNA13 Mouse Polyclonal Antibody

Supplier: Abnova
H00010672-A01
10557-616EA 243 USD
10557-616
Anti-GNA13 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GNA13 (guanine nucleotide binding protein (G protein), alpha 13)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR