Anti-CGRRF1 Mouse Monoclonal Antibody [clone: 1A4]

Supplier: Abnova
H00010668-M04
10582-090EA 335 USD
10582-090
Anti-CGRRF1 Mouse Monoclonal Antibody [clone: 1A4]
Antibodies
Type: Primary
Antigen: CGRRF1
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR