Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KLF1 Mouse Monoclonal Antibody [clone: 5G12]

Supplier: Abnova

H00010661-M04
10591-174EA 357.81 USD InStock
10591-174
Anti-KLF1 Mouse Monoclonal Antibody [clone: 5G12]
Antibodies
Type: Primary
Antigen: KLF1
Clonality: Monoclonal
Clone: 5G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty