Anti-KLF1 Mouse Monoclonal Antibody [clone: 5D7]

Supplier: Abnova
H00010661-M01
10591-168EA 335 USD
10591-168
Anti-KLF1 Mouse Monoclonal Antibody [clone: 5D7]
Antibodies
Type: Primary
Antigen: KLF1
Clonality: Monoclonal
Clone: 5D7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR