Anti-LBX1 Mouse Monoclonal Antibody [clone: 2B2]

Supplier: Abnova
H00010660-M01
10592-280EA 335 USD
10592-280
Anti-LBX1 Mouse Monoclonal Antibody [clone: 2B2]
Antibodies
Type: Primary
Antigen: LBX1
Clonality: Monoclonal
Clone: 2B2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR