Anti-KHDRBS1 Mouse Monoclonal Antibody [clone: 1A4]

Supplier: Abnova
H00010657-M03
10590-992EA 335 USD
10590-992
Anti-KHDRBS1 Mouse Monoclonal Antibody [clone: 1A4]
Antibodies
Type: Primary
Antigen: KHDRBS1
Clonality: Monoclonal
Clone: 1A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR