Anti-IGF2BP2 Mouse Monoclonal Antibody [clone: 4C6]

Supplier: Abnova
H00010644-M01
10590-426EA 335 USD
10590-426
Anti-IGF2BP2 Mouse Monoclonal Antibody [clone: 4C6]
Antibodies
Type: Primary
Antigen: IGF2BP2
Clonality: Monoclonal
Clone: 4C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR