Anti-GAS2L1 Mouse Polyclonal Antibody

Supplier: Abnova
H00010634-B01P
10557-352EA 335 USD
10557-352
Anti-GAS2L1 Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GAS2L1 (growth arrest-specific 2 like 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR