Anti-TRIM16 Mouse Monoclonal Antibody [clone: 5F4]

Supplier: Abnova
H00010626-M01
10604-612EA 335 USD
10604-612
Anti-TRIM16 Mouse Monoclonal Antibody [clone: 5F4]
Antibodies
Type: Primary
Antigen: TRIM16
Clonality: Monoclonal
Clone: 5F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR