Anti-POLR3G Mouse Polyclonal Antibody

Supplier: Abnova
H00010622-B01
10568-050EA 335 USD
10568-050
Anti-POLR3G Mouse Polyclonal Antibody
Antibodies
Type: Primary
Antigen: POLR3G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR