Anti-SPAG5 Mouse Monoclonal Antibody [clone: 5F9]

Supplier: Abnova
H00010615-M01A
10625-150EA 335 USD
10625-150
Anti-SPAG5 Mouse Monoclonal Antibody [clone: 5F9]
Antibodies
Type: Primary
Antigen: SPAG5
Clonality: Monoclonal
Clone: 5F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR