Anti-DPYSL4 Mouse Monoclonal Antibody [clone: 1F5]

Supplier: Abnova
H00010570-M01
10584-328EA 335 USD
10584-328
Anti-DPYSL4 Mouse Monoclonal Antibody [clone: 1F5]
Antibodies
Type: Primary
Antigen: DPYSL4
Clonality: Monoclonal
Clone: 1F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR