Anti-HTATIP2 Mouse Monoclonal Antibody [clone: 4G8]

Supplier: Abnova
H00010553-M01
10589-554EA 335 USD
10589-554
Anti-HTATIP2 Mouse Monoclonal Antibody [clone: 4G8]
Antibodies
Type: Primary
Antigen: HTATIP2
Clonality: Monoclonal
Clone: 4G8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR